missing translation for 'onlineSavingsMsg'
Learn More

SBI-0206965, MedChemExpress

Product Code. 16427206
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427206

Brand: Medchem Express HY16966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SBI-0206965 is a potent, selective and cell permeable autophagy kinase ULK1 inhibitor with IC50s of 108 nM for ULK1 kinase and 711 nM for the highly related kinase ULK2.

SBI-0206965 is a potent, selective and cell permeable autophagy kinase ULK1 inhibitor with IC50s of 108 nM for ULK1 kinase and 711 nM for the highly related kinase ULK2.

Specifications

Chemical Name or Material SBI-0206965
CAS 1884220-36-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21BrN4O5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (204.37 mM)
SMILES CNC(C1=CC=CC=C1OC2=NC(NC3=CC(OC)=C(OC)C(OC)=C3)=NC=C2Br)=O
Molecular Weight (g/mol) 489.32
Formula Weight 489.32
Percent Purity 98.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.