missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SBNO1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17941801
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941801 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941801 Supplier Invitrogen™ Supplier No. PA5144033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human Raji whole cell. IHC: human appendiceal adenocarcinoma tissue, human breast cancer tissue, human esophageal squamous carcinoma tissue, human gall bladder adenosquamous carcinoma tissue, human lung cancer tissue, human placenta tissue, mouse testis tissue, rat testis tissue. ICC/IF: A549 cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

SBNO1, also named as Monocyte protein 3 or MOP3, is a 1393 amino acid protein, which belongs to the SBNO family. SBNO1 play an important role in development of the vertebrate central nervous system. SBNO1 has four isoforms with MW 154 kDa and 80 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBNO1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SBNO1
Gene Accession No. A3KN83, Q5BJL5, Q689Z5
Gene Alias 9330180L10Rik; AI849128; AW557836; BC021875; Monocyte protein 3; MOP3; MOP-3; mSno1; Protein strawberry notch homolog 1; Sbno; SBNO1; Sno; sno, strawberry notch homolog 1; sno, strawberry notch homolog 1 (Drosophila); strawberry notch homolog 1; strawberry notch homolog 1 (Drosophila)
Gene Symbols SBNO1
Host Species Rabbit
Immunogen E.coli-derived human SBNO1 recombinant protein (Position: Q43-E1303).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243272, 304470, 55206
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.