missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SBSN Antibody (SBSN/7963), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23983FR
This item is not returnable.
View return policy
Description
SBSN Monoclonal antibody specifically detects SBSN in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SBSN | |
| Monoclonal | |
| DyLight 680 | |
| HLAR698, MGC75533, suprabasin, UNQ698 | |
| Recombinant fragment of human SBSN protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SBSN/7963 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 374897 | |
| Store at 4°C in the dark. | |
| IgG2 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction