missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SC35 Antibody (SR2484), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27758
This item is not returnable.
View return policy
Description
SC35 Monoclonal antibody specifically detects SC35 in Human,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| SC35 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6427 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SR2484 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:20 | |
| PR264, SC35, SC-35, serine/arginine-rich splicing factor 2, SFRS2, SFRS2A, splicing component, 35 kDa, splicing factor SC35, splicing factor, arginine/serine-rich 2, SR splicing factor 2, SRp30b | |
| A synthetic peptide of human SC35 (Uniprot #: Q01130) | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction