missing translation for 'onlineSavingsMsg'
Learn More

SC66, MedChemExpress

Product Code. 16443877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443877 50 mg 50mg
16480398 5 mg 5mg
16447938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16443877 Supplier Medchem Express Supplier No. HY19832
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SC66 is an Akt inhibitor, reduces cell viability in a dose- and time-dependent manner, inhibits colony formation and induces apoptosis in hepatocellular carcinoma (HCC) cells.

SC66 is an Akt inhibitor, reduces cell viability in a dose- and time-dependent manner, inhibits colony formation and induces apoptosis in hepatocellular carcinoma (HCC) cells.

Specifications

Chemical Name or Material SC66
Physical Form Solid
CAS 871361-88-5
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H16N2O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 25 mg/mL (90.47 mM; Need ultrasonic)
SMILES O=C1/C(CCC/C1=C\C2=CC=NC=C2)=C/C3=CC=NC=C3
Molecular Weight (g/mol) 276.33
Formula Weight 276.33
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.