missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sca-1/Ly6 Antibody (177228), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1226K
This item is not returnable.
View return policy
Description
Sca-1/Ly6 Monoclonal antibody specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Sca-1/Ly6 | |
| Monoclonal | |
| DyLight 488 | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele, Leu27-Gly119, Accession # CAA28351 | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 177228 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58530 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction