missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCAMP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11548571
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11548571 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11548571 Supplier Invitrogen™ Supplier No. PA1739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-739 detects secretory carrier membrane protein 1 (SCAMP 1) from human, mouse and rat samples. PA1-739 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~31 kDa protein representing SCAMP 1 in rat and mouse brain tissues as well as HeLa cell lysates. PA1-739 immunizing peptide corresponds to amino acid residues 1-17 from human SCAMP 1. This sequence is completely conserved between human, rat, and porcine SCAMP 1. PA1-739 immunizing peptide (Cat. # PEP-105) is available for use in neutralization and control experiments.

Secretory carrier membrane proteins (SCAMPs) are proteins that are components of post-Golgi membranes. These ubiquitously expressed proteins are implicated to function in membrane trafficking. In fibroblasts, SCAMPs are concentrated in compartments that are involved in the recycling of cell surface receptors and endocytosis. In neurons, SCAMPs are associated with synaptic vesicles, secretion granules and transporter vesicles. Of the three known SCAMPs, SCAMP 1 is the most abundant. SCAMP 1 has been shown to be required for the full execution of stable exocytosis in mast cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCAMP1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene SCAMP1
Gene Accession No. O15126, P56603, Q8K021
Gene Alias 4930505M11Rik; AI415563; AW122395; SCAMP; SCAMP 37; SCAMP1; SCAMP37; Secretory carrier membrane protein 1; secretory carrier-associated membrane protein 1
Gene Symbols SCAMP1
Host Species Rabbit
Immunogen Synthetic Peptide: M(1) S D F D S N P F A D P D L N N P(17) C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107767, 29521, 9522
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.