missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94890-0.1ml
This item is not returnable.
View return policy
Description
SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human samples. It is validated for Western Blot
Specifications
| SCAMP2 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-152 of human SCAMP2 (NP_005688.2). MSAFDTNPFADPVDVNPFQDPSVTQLTNAPQGGLAEFNPFSETNAATTVPVTQLPGSSQPAVLQPSVEPTQPTPQAVVSAAQAGLLRQQEELDRKAAELERKERELQNTVANLHVRQNNWPPLPSWCPVKPCFYQDFSTEIPADYQRICKML | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 10066 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu