missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94890-0.02ml
This item is not returnable.
View return policy
Description
SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human samples. It is validated for Western Blot
Specifications
| SCAMP2 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-152 of human SCAMP2 (NP_005688.2). MSAFDTNPFADPVDVNPFQDPSVTQLTNAPQGGLAEFNPFSETNAATTVPVTQLPGSSQPAVLQPSVEPTQPTPQAVVSAAQAGLLRQQEELDRKAAELERKERELQNTVANLHVRQNNWPPLPSWCPVKPCFYQDFSTEIPADYQRICKML | |
| 0.02 mL | |
| Membrane Trafficking and Chaperones | |
| 10066 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction