Learn More
SCAMP2 Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506956S
Encompass_Preferred
 View more versions of this product
Product Code. 17576652
  • € 242.00 / 30µL
Description
SCAMP2 Monoclonal antibody specifically detects SCAMP2 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
SCAMP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13651
Mouse
Affinity chromatography
RUO
3587
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Immunocytochemistry, Western Blot
OTI2F3
Unconjugated
Il10ra
AI875466; I79_023583; Sc2; scamp 2; SCAMP2; Secretory carrier membrane protein 2; secretory carrier-associated membrane protein 2; testis secretory sperm-binding protein Li 219p
Full length human recombinant protein of human IL10RA produced in HEK293T cell.