Learn More
Invitrogen™ SCAP Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115869
Product Code. 17218653
  • € 506.00 / 100µL
Description
Antibody detects endogenous levels of total SCAP.
The protein encoded by this gene interacts with thyroid hormone receptor in a ligand-dependent manner and enhances thyroid hormone-dependent activation from thyroid response elements. This protein contains a bromodomain and is thought to be a nuclear receptor coactivator. Three alternatively spliced transcript variants that encode distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCAP
Polyclonal
Unconjugated
SCAP
9530044G19; EMBL:AAI31853.1}; KIAA0199; mKIAA0199; PSEC0227; SCAP; scap {ECO:0000312; Sreb cleavage-activating protein; SREBF chaperone; SREBP cleavage activating protein; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; sterol regulatory element-binding protein cleavage-activating protein
Rabbit
Affinity chromatography
RUO
22937, 235623, 301024
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
A2RRU4, Q12770, Q6GQT6
SCAP
A synthesized peptide derived from human SCAP(Accession Q12770), corresponding to amino acid residues K646-G696.
100 μL
Primary
Human, Mouse, Rat