missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCARA3 Polyclonal Antibody

Product Code. 15957824
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15957824 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15957824 Supplier Invitrogen™ Supplier No. PA565053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LDNISSFLDDH EENMHDLQYH THYAQNRTVE RFESLEGRMA SHEIEIGTIF TNINATDNHV HSMLKYLDDV RLSCTLGFHT HAEELYYLNK SVSIMLGTTD LLRERFSLLS ARLDLNVRNL SMIV Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

Seems to protect cells by scavenging oxidative molecules or harmful products of oxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARA3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCARA3
Gene Accession No. Q6AZY7
Gene Alias APC7; C130058N24Rik; cellular stress response gene protein; cellular stress response protein; CSR; CSR1; macrophage scavenger receptor-like 1; MSLR1; MSRL1; Scara3; Scavenger receptor class A member 3; scavenger receptor class A, member 3
Gene Symbols SCARA3
Host Species Rabbit
Immunogen Recombinant Human SCARA3. Recombinant protein control fragment (Product #RP-105816).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.