missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCARA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104119
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SCARA3.
Seems to protect cells by scavenging oxidative molecules or harmful products of oxidation.
Specifications
| SCARA3 | |
| Polyclonal | |
| Unconjugated | |
| SCARA3 | |
| APC7; C130058N24Rik; cellular stress response gene protein; cellular stress response protein; CSR; CSR1; macrophage scavenger receptor-like 1; MSLR1; MSRL1; Scara3; Scavenger receptor class A member 3; scavenger receptor class A, member 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 364405, 51435 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AZY7 | |
| SCARA3 | |
| A synthesized peptide derived from human SCARA3(Accession Q6AZY7), corresponding to amino acid residues A504-S554. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction