missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARA5 Antibody (673527), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4900F
This item is not returnable.
View return policy
Description
SCARA5 Monoclonal antibody specifically detects SCARA5 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| SCARA5 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 286133 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 673527 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), testis expressed scavenger receptor, UNQ2938/PRO28700 | |
| NS0 mouse myeloma cell line transfected with human SCARA5, Arg83-His495, Accession # Q6ZMJ2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction