missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARA5 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77050F
This item is not returnable.
View return policy
Description
SCARA5 Polyclonal antibody specifically detects SCARA5 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SCARA5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 286133 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), testis expressed scavenger receptor, UNQ2938/PRO28700 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SCARA5. The immunogen is located within amino acids 350 - 400 of SCARA5. Amino Acid Squence: KGDRGPKGEKGEKGDRA | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction