missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCARA5 Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.
Specifications
Specifications
| Antigen | SCARA5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | SCARA5 |
| Gene Accession No. | Q6ZMJ2 |
| Gene Alias | 4932433F15Rik; 4933425F03Rik; AV278087; NET33; SCARA5; Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); scavenger receptor hlg; Tesr; testis expressed scavenger receptor; UNQ2938/PRO28700 |
| Gene Symbols | SCARA5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?