missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARB1 Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507181S
This item is not returnable.
View return policy
Description
SCARB1 Monoclonal antibody specifically detects SCARB1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| SCARB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P25963 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4792 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| NFKBIA | |
| AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII | |
| Full length human recombinant protein of human NFKBIA produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction