missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARB1 Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507181
This item is not returnable.
View return policy
Description
Receptor for different ligands such as phospholipids, cholesterol ester, lipoproteins, phosphatidylserine and apoptotic cells. Probable receptor for HDL, located in particular region of the plasma membrane, called caveolae. Facilitates the flux of free and esterified cholesterol between the cell surface and extracellular donors and acceptors, such as HDL and to a lesser extent, apoB-containing lipoproteins and modified lipoproteins. Probably involved in the phagocytosis of apoptotic cells, via its phosphatidylserine binding activity. Receptor for hepatitis C virus glycoprotein E2. Binding between SCARB1 and E2 was found to be independent of the genotype of the viral isolate. Plays an important role in the uptake of HDL cholesteryl ester.Specifications
| SCARB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WTV0 | |
| SCARB1 | |
| Full length human recombit protein of human SCARB1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| SCARB1 | |
| CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 949 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction