missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13702-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| SCARF1 | |
| Polyclonal | |
| Unconjugated | |
| SCARF1 | |
| Acetyl LDL receptor, KIAA0149, SCARF1, SREC, SREC-I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303313, 380713, 8578 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14162, Q5ND28 | |
| Scarf1 | |
| SCARF1 Fusion Protein Ag4621 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction