missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD-1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26393FR
This item is not returnable.
View return policy
Description
SCD-1 Polyclonal antibody specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SCD-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 680 | |
| FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
| Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767] | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
| 6319 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction