missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCF/c-kit Ligand Antibody (004), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89504PE
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Monoclonal antibody specifically detects SCF/c-kit Ligand in Human samples. It is validated for ELISA
Specifications
| SCF/c-kit Ligand | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 004 | |
| ELISA | |
| DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SCF/c-kit Ligand (Uniprot#: P21583-1; Met1-Ala189). | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 4254 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction