missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCF/c-kit Ligand Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77030F
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal antibody specifically detects SCF/c-kit Ligand in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SCF/c-kit Ligand | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
| Antibody was raised against a peptide corresponding to 18 amino acids near the center of human SCF/c-kit Ligand. The immunogen is located within amino acids 100-150 of SCF/c-kit Ligand. Amino Acid Sequence: KENSSKDLKKSFKSPEPR | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 4254 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction