missing translation for 'onlineSavingsMsg'
Learn More

PeproTech SCF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Product Code. 30054694
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30054694 500 μg 500µg
17854923 25 μg 25µg
17815823 50 μg 50µg
17825633 2 x 500 μg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30054694 Supplier PeproTech Supplier No. 500P202BT500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat SCF. Anti-Rat SCF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect rat SCF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat SCF (500-P202) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant rat SCF. Western Blot: To detect Rat SCF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat SCF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P202BT-1MG will be provided as 2 x 500 μg

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene KITLG
Gene Accession No. P21581
Gene Alias 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Gene Symbols KITLG
Host Species Rabbit
Immunogen E.coli-derived, 18.4 kDa Recombinant Rat SCF.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60427
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.