Learn More
SCFD1 Antibody (OTI5G4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74004AF647
Product Code. 18168865
  • € 751.00 / 0.10mL
Description
SCFD1 Monoclonal specifically detects SCFD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCFD1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
SCFD1
Full length recombinant protein of human SCFD1 (NP_057190) produced in HEK293T cells.
0.1 mL
23256.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI5G4
Western Blot, Immunohistochemistry
C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Canine, Primate, Monkey