missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD1 Antibody (OTI5G4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74004H
This item is not returnable.
View return policy
Description
SCFD1 Monoclonal specifically detects SCFD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SCFD1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| SCFD1 | |
| Full length recombinant protein of human SCFD1 (NP_057190) produced in HEK293T cells. | |
| 0.1 mL | |
| 23256.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5G4 | |
| Western Blot, Immunohistochemistry | |
| C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction