Learn More
SCFD1 Monoclonal Antibody (OTI2E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504662
Encompass_Preferred
 View more versions of this product
Product Code. 17532152
  • € 540.87 / 100µL
Description
SCFD1 is involved in vesicular transport between the endoplasmic reticulum and the Golgi.
Specifications
Specifications
SCFD1
Monoclonal
0.87 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q62991, Q8WVM8
SCFD1
Full length human recombit protein of human SCFD1 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2E7
Unconjugated
SCFD1
C14orf163, RA410, SLY1, SLY1P, STXBP1L2
Mouse
Affinity Chromatography
RUO
23256, 480281, 54350
-20° C, Avoid Freeze/Thaw Cycles