missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD1 Monoclonal Antibody (OTI5G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504721S
This item is not returnable.
View return policy
Description
SCFD1 Monoclonal antibody specifically detects SCFD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| SCFD1 | |
| Monoclonal | |
| 0.86 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WVM8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23256 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G4 | |
| Unconjugated | |
| SCFD1 | |
| 3110021P21Rik; C14orf163; FKSG23; KIAA0917; Ra410; rSly1; Scfd1; Sec1 family domain containing 1; sec1 family domain-containing protein 1; Sly1; SLY1 homolog; Sly1p; STXBP1L2; Syntaxin-binding protein 1-like 2; vesicle transport-related protein; vesicle transport-related protein Ra410 | |
| Full length human recombinant protein of human SCFD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction