Learn More
SCGBL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21142-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| SCGBL | |
| Polyclonal | |
| Unconjugated | |
| SCGB2B2 | |
| SCGBL, secretoglobin like, Secretoglobin like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284402 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4G0G5 | |
| SCGB2B2 | |
| SCGBL Fusion Protein Ag15610 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |