missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGF/CLEC11a Antibody (MM0533-6D5), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11874MFV450
This item is not returnable.
View return policy
Description
SCGF/CLEC11a Monoclonal antibody specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SCGF/CLEC11a | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6320 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0533-6D5 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CLECSF3, LSLCL, P47, SCGF | |
| Human recombinant SCGF/CLEC11a | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction