missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCGN Monoclonal Antibody (CL0271)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531337
This item is not returnable.
View return policy
Description
Immunogen sequence: RDLFLHHKKA ISEAKLEEYT GTMMKIFDRN KDGRLDLNDL ARILALQENF LLQFKMDACS TEERKRDFEK IFAYYDVSKT GALEGPEVDG FVKDMMELVQ PSISGVDLDK FREILLRHCD VNKDGKIQKS ELALCLGLK Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
Specifications
| SCGN | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76038 | |
| SCGN | |
| Recombinant protein fragment of Human SCGN (Product #RP-102381) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL0271 | |
| Unconjugated | |
| SCGN | |
| CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin | |
| Mouse | |
| Protein A | |
| RUO | |
| 10590 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction