missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCGN Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 500.00 - € 1736.00
Specifications
| Antigen | SCGN |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17835247
|
Invitrogen™
PA5143521 |
500 μL |
€ 1736.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17836417
![]() |
Invitrogen™
PA5143619 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCGN Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.Specifications
| SCGN | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| SCGN | |
| CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin | |
| SCGN | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| O76038, Q6R556, Q91WD9 | |
| 10590, 214189, 306942 | |
| Full-length recombinant human secretagogin protein expressed in and purified from E. coli. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title