missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SCH 58261
GREENER_CHOICE

Product Code. 18024182
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024182

Brand: Tocris Bioscience™ 2270/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, highly selective A2A antagonist

SCH 58261 is a potent and selective A2A adenosine receptor competitive antagonist (Ki = 1.3 nM). Displays 323-, 53- and 100-fold selectivity over A1, A2B and A3 receptors, respectively. Reduces melanoma tumor growth and TGF-β levels in WT and P2X7R-null mice.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160098-96-4
Molecular Formula C18H15N7O

Specifications

Quantity 50 mg
Formula Weight 345.36
Purity >99%
Molecular Weight (g/mol) 345.366
Chemical Name or Material SCH 58261
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.