Learn More
Kimble™ Shear Precision Melting Tubes
Product Code. 10624491
Change view
Click to view available options
Length (Metric):
70 mm
80 mm
100 mm
Width (Metric):
0.5 mm
1 mm
1.5 mm
Unit Size:
Each
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
10624491 100 mm 1 mm Pack of 100
10172340 70 mm 1 mm Pack of 100
10580381 70 mm 1.5 mm Each
10090980 80 mm 0.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10624491 Supplier Kimble™ Supplier No. ZSCHR119162SP
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Use Kimble™ Shear Precision Melting Tubes for your laboratory melting apparatus needs.
  • Clear
  • Heat tolerant
Specifications
Product Type Melting Point Tube
Length (Metric) 100 mm
Width (Metric) 1 mm