missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCHIP1 Antibody (OTI1C8), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74008DL594
This item is not returnable.
View return policy
Description
SCHIP1 Monoclonal antibody specifically detects SCHIP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| SCHIP1 | |
| Monoclonal | |
| DyLight 594 | |
| FLJ39160, SCHIP-1schwannomin-interacting protein 1, schwannomin interacting protein 1 | |
| Full length recombinant protein of human SCHIP1 (NP_055390) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1C8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 29970 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction