missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS11 Monoclonal Antibody (OTI2D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802879S
This item is not returnable.
View return policy
Description
MRPS11 Monoclonal antibody specifically detects MRPS11 in Human samples. It is validated for Western BlotSpecifications
| MRPS11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P82912 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 64963 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2D1 | |
| Unconjugated | |
| MRPS11 | |
| 0710005I03Rik; 28S ribosomal protein S11, mitochondrial; C79873; Cervical cancer proto-oncogene 2 protein; HCC2; HCC-2; mitochondrial ribosomal protein S11; Mitochondrial small ribosomal subunit protein uS11m; MRPS11; MRP-S11; RGD1559901; RPMS11; S11mt | |
| Full length human recombinant protein of human MRPS11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction