missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSNK1G2 Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802864
This item is not returnable.
View return policy
Description
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.Specifications
| CSNK1G2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78368 | |
| CSNK1G2 | |
| Human recombit protein fragment corresponding to amino acids 163-415 of human CSNK1G2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2F2 | |
| Unconjugated | |
| CSNK1G2 | |
| CK1g2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1455 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction