missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schistosoma Mansoni Antibody (BGN/1A7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64566UV
This item is not returnable.
View return policy
Description
Schistosoma Mansoni Monoclonal specifically detects Schistosoma Mansoni in Invertebrate samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Schistosoma Mansoni | |
| Monoclonal | |
| DyLight 350 | |
| S. mansoni | |
| Native S. mansoni cercariae. | |
| 0.1 mL | |
| Primary | |
| Invertebrate | |
| Purified |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| BGN/1A7 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Reacts with antigen present in the urine of patients with Schistosomiasis. Reacts with a TCA soluble adult worm antigen in IF/ELISA. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction