Learn More
Schuett Biotec™ Schuett Phoenix II Standard Burner
Brand: Schuett Biotec™ 3355202
Additional Details : Weight : 0.99000kg
Specifications
Standard Burner | |
Flaming or annealing, Heating, Sterilizing | |
1 Ea. |
Natural Gas, Butane, Propane | |
85 mm | |
0.99 kg |