missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCIMP Antibody (NVL-07), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22349DL594
This item is not returnable.
View return policy
Description
SCIMP Monoclonal antibody specifically detects SCIMP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Intracellular), CyTOF
Specifications
| SCIMP | |
| Monoclonal | |
| DyLight 594 | |
| transmembrane protein C17orf87, C17orf87, chromosome 17 open reading frame 87, DTFT5783, FLJ32580, SLP adapter and CSK-interacting membrane protein, SLP adaptor and CSK interacting membrane protein, SLP65/SLP76, Csk-interacting membrane protein, UNQ5783 | |
| Recombinant intracellular part of human SCIMP. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow Cytometry (Intracellular Staining) | |
| NVL-07 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 388325 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction