missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCIN Antibody (S07-6G2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 420.00 - € 642.60
Specifications
| Antigen | SCIN |
|---|---|
| Clone | S07-6G2 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617583
|
Novus Biologicals
NBP3-19768-100ul |
100 μg |
€ 680.00 € 642.60 / 100µL Save € 37.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628793
|
Novus Biologicals
NBP3-19768-50ul |
50 μg |
€ 444.00 € 420.00 / 50µL Save € 24.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCIN Monoclonal antibody specifically detects SCIN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SCIN | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 85477 | |
| IgG | |
| Affinity purified |
| S07-6G2 | |
| Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| KIAA1905adseverin, Scinderin | |
| A synthetic peptide of human SCIN (Uniprot # Q9Y6U3) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title