missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCIN Recombinant Rabbit Monoclonal Antibody (23GB4135), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | SCIN |
|---|---|
| Clone | 23GB4135 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088127
![]() |
Invitrogen™
MA552658 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087479
|
Invitrogen™
MA552659 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCIN Recombinant Monoclonal antibody specifically detects SCIN in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SCIN | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q60604, Q9Y6U3 | |
| 20259, 298975, 85477 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4135 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Adseverin; AW545522; Gelsolin-like protein; KIAA1905; SCIN; Scind; Scinderin | |
| A synthesized peptide derived from human SCIN (650-715AA). | |
| Primary | |
| -20°C | |
| SCIN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title