missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCL/Tal1 Antibody (BTL73), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08616JF549
This item is not returnable.
View return policy
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| SCL/Tal1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| Recombinant human full-length TAL1 protein (Uniprot: P17542) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| BTL73 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6886 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction