missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCL/Tal1 Mouse anti-Human, Clone: BTL73, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | SCL/Tal1 |
|---|---|
| Clone | BTL73 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, ELISA |
| Applications | Western Blot, ELISA |
Description
SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| SCL/Tal1 | |
| 0.2 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 6886 | |
| IgG1 κ | |
| Protein A or G purified |
| BTL73 | |
| Western Blot 1 to 2 μg/mL, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| Recombinant human full-length SCL/Tal1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title