missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ICAM-5 hIgG-His Protein

Product Code. 18607736 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
50 μg
500 μg
Unit Size:
100µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18607736

Brand: Novus Biologicals™ NBP30577450ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) and 10% glycerol
Gene ID (Entrez) 7087
Molecular Weight (g/mol) MolecularWeight-theroretical: 113.8 kDa
Quantity 50 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than 1 EU / 1 μg of protein (determined by LAL method)
Gene Alias ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Gene Symbol ICAM5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Immunology, Neuroscience
Purity or Quality Grade >90%, by SDS-PAGE
Protein ICAM-5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.