missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtIP Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77165R
This item is not returnable.
View return policy
Description
CtIP Polyclonal antibody specifically detects CtIP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CtIP | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human RBBP8. The immunogen is located within amino acids 430 - 480 of RBBP8. Amino Acid Squence: CRTSKRKKTEEESEH | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 5932 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction