missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCN3A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16617174
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16617174 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16617174 Supplier Invitrogen™ Supplier No. PA5100688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCN3A.

Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Scn3a
Gene Accession No. Q9NY46
Gene Alias BIII; brain III voltage-gated sodium channel; Gm1000; KIAA1356; NAC3; Nav1.3; SCIII; Scn2a; Scn3a; Sodium channel protein brain III subunit alpha; sodium channel protein type 3 subunit alpha; Sodium channel protein type III subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type III, alpha; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type III, alpha subunit; sodium voltage-gated channel alpha subunit 3; voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.3
Gene Symbols Scn3a
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCN3A(Accession Q9NY46), corresponding to amino acid residues L491-M541.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6328
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.