Learn More
Invitrogen™ SCN3B Monoclonal Antibody (N396/29), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545738
Product Code. 17866002
  • € 455.00 / 100µg
Description
A 1:100 dilution of MA5-45738 was sufficient for detection of NavBeta3 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4. This antibody was formerly sold as clone S396-29.
Voltage-gated sodium channels are transmembrane glycoprotein complexes composed of a large alpha subunit and one or more regulatory beta subunits. They are responsible for the generation and propagation of action potentials in neurons and muscle. This gene encodes one member of the sodium channel beta subunit gene family, and influences the inactivation kinetics of the sodium channel. Two alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCN3B
Monoclonal
1 mg/mL
PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Q8BHK2, Q9JK00, Q9NY72
Scn3b
Fusion protein amino acids 1-215 (full-length) of rat NavBeta3.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Western Blot, Immunocytochemistry
N396/29
FITC
Scn3b
1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit
Mouse
Protein G
RUO
235281, 245956, 55800
4°C, store in dark