missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCO1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97289V
This item is not returnable.
View return policy
Description
SCO1 Polyclonal antibody specifically detects SCO1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SCO1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| protein SCO1 homolog, mitochondrial, SCO cytochrome oxidase deficient homolog 1 (yeast), SCOD1SCO (cytochrome oxidase deficient, yeast) homolog 1 | |
| Produced in rabbits immunized with E. coli-derived Human SCO1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 6341 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction