missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95256-0.1ml
This item is not returnable.
View return policy
Description
SCP1 Polyclonal antibody specifically detects SCP1 in Mouse samples. It is validated for Western Blot
Specifications
| SCP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human SYCP1 (NP_003167.2). EGLSRVYSKLYKEAEKIKKWKVSTEAELRQKESKLQENRKIIEAQRKAIQELQFGNEKVSLKLEEGIQENKDLIKENNATRHLCNLLKETCARSAEKTKKYEYEREETRQVYMDLNNNIEKMITAFEELRVQAENSRLEMHFKLKEDYEKIQHLEQEYKKEINDKEKQVSLLLIQITEKENKMKDLTFLLEESRDKVNQLEEKTKLQSENLKQSIEKQHHLTKELEDIKVSLQRSVSTQKALEEDLQIATKTICQLTEEKETQMEESNKARAAHSFVVTEFETTVCSLEELLRTEQQRLEK | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6847 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction