missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB300-228JF669
This item is not returnable.
View return policy
Description
SCP1 Polyclonal antibody specifically detects SCP1 in Human,Mouse,Primate,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunoblot,Simple Western,Immunocytochemistry/ Immunofluorescence
Specifications
| SCP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoblot, Simple Western, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
| A C-terminal synthetic peptide made to the human SCP1 protein sequence. [UniProt# Q15431] | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6847 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction